Assay Details
| Gene Symbol: Tex21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025658.1 | NP_001020829.1 | 4 - 5 | 1075 | 81 | |
| XM_006240226.2 | XP_006240288.1 | 6 - 7 | 1278 | 81 | ||
| XM_006240227.2 | XP_006240289.1 | 6 - 7 | 1322 | 81 | ||
| XM_008764743.1 | XP_008762965.1 | 6 - 7 | 1316 | 81 | ||
| XM_008764744.1 | XP_008762966.1 | 5 - 6 | 1193 | 81 | ||
| XM_008764745.1 | XP_008762967.1 | 4 - 5 | 798 | 81 | ||
| GenBank mRNA | BC097422.1 | - | 4 - 5 | 1075 | 7454996 | 81 |
More Information
Updated Mapping Information
Set Membership: |
|

Tex21
Genomic Map