Assay Details
| Gene Symbol: Proca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025758.1 | NP_001020929.1 | 1 - 2 | 223 | 58 | |
| GenBank mRNA | BC098036.1 | - | 1 - 2 | 223 | 7456533 | 58 |
More Information
Updated Mapping Information
Set Membership: |
|

Proca1
Genomic Map