Assay Details
| Gene Symbol: Chst12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037775.2 | NP_001032864.1 | 1 | 1236 | 106 | |
| XM_006248935.1 | XP_006248997.1 | 2 | 1330 | 106 | ||
| XM_006248936.2 | XP_006248998.1 | 2 | 2379 | 106 | ||
| XM_006248937.2 | XP_006248999.1 | 2 | 3117 | 106 | ||
| XM_008768971.1 | XP_008767193.1 | 3 | 2261 | 106 | ||
| GenBank mRNA | BC100096.1 | - | 1 | 1240 | 7375118 | 106 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chst12
Genomic Map