Assay Details
| Gene Symbol: Gpr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109096.1 | NP_001102566.1 | 1 | 1183 | 83 | |
| XM_006251616.2 | XP_006251678.1 | 1 | 1288 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr75
Genomic Map