Assay Details
| Gene Symbol: Klhl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107944.1 | NP_001101414.1 | 1 | 649 | 83 | |
| GenBank mRNA | BC168717.1 | - | 1 | 632 | 9102826 | 83 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Klhl9
Genomic Map