Assay Details
| Gene Symbol: Hoxa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109233.2 | NP_001102703.2 | 476 | 53 | ||
| XM_006236501.2 | XP_006236563.1 | 1885 | 53 | |||
| XM_006236503.2 | XP_006236565.1 | 1906 | 53 | |||
| XM_008762907.1 | XP_008761129.1 | 1133 | 53 | |||
| Gene Symbol: LOC100911622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752181.3 | XP_003752229.1 | 2 - 3 | 1885 | 53 | |
| XM_006257702.2 | XP_006257764.1 | 2 - 3 | 1906 | 53 | ||
| XM_008773710.1 | XP_008771932.1 | 1 - 2 | 2149 | 53 | ||
| XM_008773711.1 | XP_008771933.1 | 2 - 3 | 1133 | 53 | ||
| Gene Symbol: NEWGENE_1587253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762940.1 | XP_008761162.1 | 1 - 2 | 2149 | 53 | |
| XM_008762941.1 | XP_008761163.1 | 2 - 3 | 1906 | 53 | ||
| XM_008762942.1 | XP_008761164.1 | 2 - 3 | 1885 | 53 | ||
| XM_008762943.1 | XP_008761165.1 | 2 - 3 | 1133 | 53 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Hoxa9
Genomic Map