Assay Details
| Gene Symbol: Rnf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113748.1 | NP_001107220.1 | 1 | 1150 | 81 | |
| GenBank mRNA | BC158712.1 | - | 1 | 1150 | 9035415 | 81 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf26
Genomic Map