Assay Details
| Gene Symbol: Crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011710.2 | NP_001011710.1 | 5 - 6 | 403 | 107 | |
| XM_006244643.2 | XP_006244705.1 | 4 - 5 | 386 | 107 | ||
| GenBank mRNA | AB009662.1 | - | 4 - 5 | 266 | 107 | |
| BC078799.1 | - | 5 - 6 | 352 | 7112769 | 107 | |
| CK469282.1 | - | 5 - 6 | 403 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crisp2
Genomic Map