Assay Details
| Gene Symbol: Adnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127373.1 | NP_001120845.1 | 2 - 3 | 381 | 70 | |
| GenBank mRNA | BC166606.1 | - | 2 - 3 | 381 | 7934474 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Adnp2
Genomic Map