Assay Details
| Gene Symbol: Kcnk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134912.1 | NP_001128384.1 | 1 - 2 | 324 | 107 | |
| GenBank mRNA | EU152989.1 | - | 1 - 2 | 324 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnk18
Genomic Map