Assay Details
| Gene Symbol: Ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100728.1 | NP_001094198.1 | 1 - 2 | 220 | 85 | |
| GenBank mRNA | BC091255.1 | - | 1 - 2 | 194 | 85 | |
| CB792766.1 | - | 1 - 2 | 267 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc43
Genomic Map