Assay Details
| Gene Symbol: Apom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019373.2 | NP_062246.1 | 1 - 2 | 188 | 64 | |
| GenBank mRNA | AF207821.1 | - | 1 - 2 | 165 | 64 | |
| BC089807.1 | - | 1 - 2 | 137 | 7323885 | 64 | |
| FQ210093.1 | - | 1 - 2 | 187 | 64 | ||
| FQ219449.1 | - | 1 - 2 | 188 | 64 | ||
More Information
Set Membership: |
|

Apom
Genomic Map