Assay Details
| Gene Symbol: Ptger1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278475.1 | NP_001265404.1 | 2 - 3 | 1242 | 63 | |
| XM_008772362.1 | XP_008770584.1 | 2 - 3 | 1894 | 63 | ||
| GenBank mRNA | D88751.1 | - | 1 - 2 | 1534 | 63 | |
More Information
Set Membership: |
|

Ptger1
Genomic Map