Assay Details
| Gene Symbol: Retnla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053333.1 | NP_445785.1 | 1 - 2 | 154 | 67 | |
| XM_006248283.2 | XP_006248345.1 | 2 - 3 | 246 | 67 | ||
| GenBank mRNA | AF323085.1 | - | 1 - 2 | 154 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Retnla
Genomic Map