Assay Details
| Gene Symbol: Prelid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009636.1 | NP_001009636.1 | 1 | 48 | 96 | |
| GenBank mRNA | BC088284.1 | - | 1 | 48 | 7323653 | 96 |
| FQ212767.1 | - | 1 | 60 | 96 | ||
| FQ226295.1 | - | 1 | 62 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088284.1,NM_001009636.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prelid1
Genomic Map