Assay Details
| Gene Symbol: Ugt1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039549.1 | NP_001034638.1 | 1 | 44 | 111 | |
| GenBank mRNA | AF461734.1 | - | 1 | 53 | 111 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF461734.1,NM_001039549.1 transcripts.
Set Membership: |
|

Ugt1a5
Genomic Map