Assay Details
| Gene Symbol: Lefty1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109080.1 | NP_001102550.1 | 1 | 48 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109080.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lefty1
Genomic Map