Assay Details
| Gene Symbol: Pcdhgc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164288.1 | NP_001157760.1 | 2349 | 60 | ||
| GenBank mRNA | GQ131870.1 | - | 2349 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhgc5| Gene Symbol: Pcdhgc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164288.1 | NP_001157760.1 | 2349 | 60 | ||
| GenBank mRNA | GQ131870.1 | - | 2349 | 60 | ||
Set Membership: |
|