Assay Details
| Gene Symbol: Klrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019261.1 | NP_062134.1 | 1 | 52 | 84 | |
| GenBank mRNA | AF021349.1 | - | 1 | 52 | 84 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF021349.1,NM_019261.1 transcripts.
Set Membership: |
|

Klrc2
Genomic Map