Assay Details
| Gene Symbol: Mir99b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031857.1 | 1 | 38 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 709-like | LOC102552823 | -195.6kb | Sense | Chr. 1 - 59483702 - 59508630 on NCBI Build Rnor_6.0 | |
| Downstream | microRNA mir-3596c | Mir3596c | rno-mir-3596c | 87.0 | Anti-sense | Chr. 1 - 59704372 - 59704488 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir99b
Genomic Map