Assay Details
| Gene Symbol: Mir483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032112.1 | 1 | 33 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102547221 | LOC102547221 | -78.2kb | Anti-sense | Chr. 1 - 215747170 - 215753861 on NCBI Build Rnor_6.0 | |
| Downstream | vegetative cell wall protein gp1-like | LOC102548568 | 12.5kb | Sense | Chr. 1 - 215844626 - 215847533 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir483
Genomic Map