Assay Details
| Gene Symbol: Mir290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031937.1 | 1 | 42 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 291a | Mir291a | rno-mir-291a | -217.0 | Sense | Chr. 1 - 64538407 - 64538488 on NCBI Build Rnor_6.0 |
| Downstream |
similar to solute carrier family 7 (cationic amino acid transporter, y+ system), member 3 |
LOC292543 | 79.6kb | Anti-sense | Chr. 1 - 64618350 - 64632132 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir290
Genomic Map