Assay Details
| Gene Symbol: Mir489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032105.1 | 1 | 52 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-653 | Mir653 | rno-mir-653 | -545.0 | Sense | Chr. 4 - 28658882 - 28658978 on NCBI Build Rnor_6.0 |
| Downstream | tissue factor pathway inhibitor 2 | Tfpi2 | 288.7kb | Sense | Chr. 4 - 28948343 - 28952765 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir489
Genomic Map