Assay Details
| Gene Symbol: Mir329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031776.1 | 1 | 42 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-758 | Mir758 | rno-mir-758 | -593.0 | Sense | Chr. 6 - 133861498 - 133861575 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 494 | Mir494 | rno-mir-494 | 2.1kb | Sense | Chr. 6 - 133864370 - 133864452 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir329
Genomic Map