Assay Details
| Gene Symbol: Mir20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031793.1 | 1 | 43 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 19a | Mir19a | rno-mir-19a | -91.0 | Sense | Chr. 15 - 100180162 - 100180243 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 19b-1 | Mir19b1 |
Mir19b-1, rno-mir-19b-1 |
46.0 | Sense | Chr. 15 - 100180464 - 100180550 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir20a
Genomic Map