Assay Details
| Gene Symbol: Mirlet7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031802.1 | 1 | 41 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA let-7c-2 | Mirlet7c-2 | rno-let-7c-2 | -321.0 | Sense | Chr. 7 - 126590212 - 126590306 on NCBI Build Rnor_6.0 |
| Downstream | peroxisome proliferator activated receptor alpha | Ppara | PPAR | 28.2kb | Sense | Chr. 7 - 126618867 - 126687282 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7b
Genomic Map