Assay Details
| Gene Symbol: Mir511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032742.1 | 1 | 32 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 236 | Tmem236 | Fam23a | -40.9kb | Sense | Chr. 17 - 81280636 - 81342110 on NCBI Build Rnor_6.0 |
| Downstream |
solute carrier family 39 (zinc transporter), member 12 |
Slc39a12 | 72.7kb | Sense | Chr. 17 - 81455753 - 81541742 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir511
Genomic Map