Assay Details
| Gene Symbol: Mir3562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037344.1 | 1 | 53 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-3594 | Mir3594 | rno-mir-3594 | -2.0kb | Sense | Chr. 10 - 109244637 - 109244701 on NCBI Build Rnor_6.0 |
| Downstream | ENTH domain containing 2 | Enthd2 | RGD1307410 | 24.3kb | Sense | Chr. 10 - 109271161 - 109278510 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3562
Genomic Map