Assay Details
| Gene Symbol: Mir3594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037395.1 | 1 | 37 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | apoptosis-associated tyrosine kinase | Aatk | -20.2kb | Sense | Chr. 10 - 109184338 - 109224482 on NCBI Build Rnor_6.0 | |
| Downstream | microRNA mir-3562 | Mir3562 | rno-mir-3562 | 2.0kb | Sense | Chr. 10 - 109246744 - 109246855 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3594
Genomic Map