Assay Details
| Gene Symbol: Mir19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031822.1 | 1 | 44 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 18a | Mir18a | rno-mir-18a | -52.0 | Sense | Chr. 15 - 100180015 - 100180110 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 20a | Mir20a | rno-mir-20a | 91.0 | Sense | Chr. 15 - 100180334 - 100180418 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir19a
Genomic Map