Assay Details
| Gene Symbol: Mir3549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037325.1 | 1 | 52 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-193 | Mir193 |
Mir193a, rno-mir-193, rno-mir-193a |
Anti-sense | Chr. 10 - 67065896 - 67065981 on NCBI Build Rnor_6.0 | |
| Downstream | microRNA 365 | Mir365 | rno-mir-365 | 13.8kb | Anti-sense | Chr. 10 - 67079796 - 67079881 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3549
Genomic Map