Assay Details
| Gene Symbol: Lrrc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107204.2 | NP_001100674.1 | 1 - 2 | 2134 | 69 | |
| XM_006250557.1 | XP_006250619.1 | 4 - 5 | 2445 | 69 | ||
| GenBank mRNA | FQ212117.1 | - | 4 - 5 | 2450 | 69 | |
| FQ217980.1 | - | 4 - 5 | 2400 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc8b
Genomic Map