Assay Details
| Gene Symbol: Mir365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031952.1 | 1 | 41 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3549 | Mir3549 | rno-mir-3549 | -13.8kb | Anti-sense | Chr. 10 - 67065885 - 67065996 on NCBI Build Rnor_6.0 |
| Downstream |
UTP6, small subunit (SSU) processome component, homolog (yeast) |
Utp6 | RGD1311017 | 175.3kb | Anti-sense | Chr. 10 - 67255177 - 67285738 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir365
Genomic Map