Assay Details
| Gene Symbol: Mir291a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031938.1 | 1 | 46 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-292 | Mir292 | rno-mir-292 | -207.0 | Sense | Chr. 1 - 64538119 - 64538200 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 290 | Mir290 | rno-mir-290 | 217.0 | Sense | Chr. 1 - 64538705 - 64538785 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir291a
Genomic Map