Assay Details
| Gene Symbol: Pate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270389.1 | NP_001257318.1 | 4 - 5 | 239 | 77 | |
| GenBank mRNA | JF412808.1 | - | 4 - 5 | 239 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pate1
Genomic Map