Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270867.1 | NP_001257796.1 | 1 - 2 | 196 | 100 | |
| NM_053601.2 | NP_446053.1 | 1 - 2 | 196 | 154 | ||
| XM_008762390.1 | XP_008760612.1 | 1 - 2 | 191 | 149 | ||
| GenBank mRNA | CB711943.1 | - | 1 - 2 | 132 | 154 | |
| FM037467.1 | - | 1 - 2 | 196 | 100 | ||
| FQ213121.1 | - | 1 - 2 | 154 | 154 | ||
| U08290.1 | - | 1 - 2 | 115 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nnat
Genomic Map