Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270867.1 | NP_001257796.1 | 2 - 3 | 222 | 100 | |
| GenBank mRNA | FM037467.1 | - | 2 - 3 | 222 | 100 | |
More Information
Set Membership: |
|

Nnat
Genomic Map