Assay Details
| Gene Symbol: Ago3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271193.1 | NP_001258122.1 | 8 - 9 | 1278 | 64 | |
| XM_008764023.1 | XP_008762245.1 | 5 - 6 | 1780 | 64 | ||
| XM_008764024.1 | XP_008762246.1 | 4 - 5 | 1744 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ago3
Genomic Map