Assay Details
| Gene Symbol: Foxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017077.2 | NP_058773.1 | 1 - 2 | 139 | 99 | |
| GenBank mRNA | L09648.1 | - | 1 - 2 | 140 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Foxa3
Genomic Map