Assay Details
| Gene Symbol: Prpmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172065.2 | NP_742062.1 | 1 - 3 | 80 | 105 | |
| GenBank mRNA | L17318.1 | - | 1 - 3 | 81 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpmp5
Genomic Map