Assay Details
| Gene Symbol: Prpmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172065.2 | NP_742062.1 | 4 - 5 | 939 | 93 | |
| GenBank mRNA | L17318.1 | - | 4 - 5 | 940 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Prpmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172065.2 | NP_742062.1 | 4 - 5 | 939 | 93 | |
| GenBank mRNA | L17318.1 | - | 4 - 5 | 940 | 93 | |
Set Membership: |
|