Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181687.2 | NP_859015.1 | 1 - 2 | 199 | 73 | |
| GenBank mRNA | BC127473.1 | - | 1 - 2 | 133 | 8364201 | 73 |
| FQ213576.1 | - | 1 - 2 | 157 | 73 | ||
| U09785.1 | - | 1 - 2 | 118 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nnat
Genomic Map