Assay Details
| Gene Symbol: Sptssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271299.1 | NP_001258228.1 | 3 | 1922 | 114 | |
| NM_001271301.1 | NP_001258230.1 | 2 | 1827 | 114 | ||
| GenBank mRNA | BC087706.1 | - | 2 | 1827 | 114 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sptssb
Genomic Map