Assay Details
| Gene Symbol: LOC100911827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110710.1 | 3 - 4 | 806 | 70 | ||
| GenBank mRNA | FQ211983.1 | - | 3 - 4 | 807 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ladybird homeobox 1 | Lbx1 | RGD1564197 | -97.8kb | Sense | Chr. 1 - 264973315 - 264975132 on NCBI Build Rnor_6.0 |
| Downstream | beta-transducin repeat containing E3 ubiquitin protein ligase | Btrc | beta-TrCP1 | 26.3kb | Anti-sense | Chr. 1 - 265100100 - 265270263 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

LOC100911827
Genomic Map