Assay Details
| Gene Symbol: Prl3d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ329282.1 | - | 5 | 817 | 100 | |
| DQ329283.1 | - | 5 | 817 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prl3d2
Genomic Map