Assay Details
| Gene Symbol: Serpinb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135867.1 | NP_001129339.1 | 4 - 5 | 421 | 87 | |
| XM_006249637.2 | XP_006249699.1 | 3 - 4 | 433 | 87 | ||
| Gene Symbol: LOC103690041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751227.3 | XP_003751275.3 | 2 - 3 | 323 | 87 | |
More Information
Set Membership: |
|

Serpinb12
Genomic Map