Assay Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033899.1 | NP_001029071.1 | 10 | 1078 | 95 | |
| GenBank mRNA | BC101870.1 | - | 10 | 1078 | 5621864 | 95 |
More Information
Set Membership: |
|

Sms
Genomic Map