Assay Details
| Gene Symbol: LOC100912475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250360.2 | XP_006250422.1 | 1 - 2 | 301 | 79 | |
| Gene Symbol: Cnst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250412.2 | XP_006250474.1 | 1 - 2 | 301 | 79 | |
| XM_006250413.2 | XP_006250475.1 | 1 - 2 | 301 | 79 | ||
| XM_573525.5 | XP_573525.5 | 1 - 2 | 301 | 79 | ||
More Information
Set Membership: |
|

LOC100912475
Genomic Map