Assay Details
| Gene Symbol: Hspa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212504.1 | NP_997669.1 | 1 | 1517 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_212504.1 transcripts.
Set Membership: |
|

Hspa1b
Genomic Map