Assay Details
| Gene Symbol: Bmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191856.2 | NP_001178785.1 | 1 | 280 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191856.2 transcripts.
Set Membership: |
|

Bmp7
Genomic Map